Order Entry
Export
ContactUsLinkComponent
Chloramphenicol
Health
Chloramphenicol
Catalog # EHERC11120000
Supplier:  EHRENSTORFER
CAS Number:  
Health
Chloramphenicol
Catalog # EHERC11120000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :323.13 g/mol
Merck Index:13,02087
Boiling Pt:645 °C (1013 hPa)
Melting Pt:150 °C
Density:1.49 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-287-4
 :GHS08
CAS Number:56-75-7
Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MDL Number:MFCD00078159