Chloramphenicol
Glass bottle for solids, 500 mg
Catalog # PROOMM0516.00
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
- CAS Number:56-75-7
- Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
- MDL Number:MFCD00078159
- Molecular Weight:323.13 g/mol
- Merck Index:13,02087
- Boiling Pt:645 °C (1013 hPa)
- Melting Pt:150 °C
- Density:1.49 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-287-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



