
Chloramphenicol
Supplier: Roth Carl
Certificates
About this item
CAS Number:56-75-7
:
Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MDL Number:MFCD00078159
:323.13 g/mol
Merck Index:13,02087
Boiling Pt:645 °C (1013 hPa)
Melting Pt:150 °C
Density:1.49 g/cm³ (20 °C)
EINECS:200-287-4
:GHS08
Storage Temperature:Ambient