Order Entry
Export
Orders LinkContactUsLinkComponent
DL-Alanine

DL-Alanine

Glass bottle for solids, 100 g
Catalog # ROTH9798.1
Supplier:   Roth Carl
DL-Alanine
Catalog # ROTH9798.1
Supplier:  Roth Carl
CAS Number:  302-72-7
DL-Alanine

Glass bottle for solids, 100 g

CAS Number:  302-72-7
Price
56,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine
  • CAS Number:302-72-7
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00064408
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00203
  • Boiling Pt:189 °C (1013 hPa)
  • Melting Pt:79 °C
  • Density:1.42 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:206-126-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications