Order Entry
Export
Orders LinkContactUsLinkComponent
 

DL-Alanine ≥99%

DL-Alanine ≥99%
Certificates
DL-Alanine ≥99%
DL-Alanine ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 302-72-7 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Melting Point: ca 292° dec. Beilstein Reference: 635807 Merck Reference: 14,204 MDL No.: MFCD00064408

Synonyms:  (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine
  • CAS Number:302-72-7
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00064408
  • Molecular Weight:89.09 g/mol
  • Merck Index:14,204
  • Boiling Pt:189 °C (1013 hPa)
  • Melting Pt:∼ 292 °C (dec.)
  • Density:1.42
  • Storage Temperature:Ambient
  • EINECS:206-126-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 49,40 €
    49,40 €
    1 * 250 g
       49,40 €

  • Glass bottle for solids
    1.000 g
    Log in for availability
    Log in for availability
    1 * 1.000 g
    1 * 1.000 g - 137,00 €
    137,00 €
    1 * 1.000 g
       137,00 €

  • Glass bottle for solids
    5.000 g
    Log in for availability
    Log in for availability
    1 * 5.000 g
    1 * 5.000 g - 522,00 €
    522,00 €
    1 * 5.000 g
       522,00 €