Order Entry
Export
ContactUsLinkComponent
2,5-Diaminopyridine dihydrochloride
2,5-Diaminopyridine dihydrochloride
Catalog # MOLE25440537-25G
Supplier:  Molekula
CAS Number:  
2,5-Diaminopyridine dihydrochloride
Catalog # MOLE25440537-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :182.05 g/mol
Melting Pt:264 °C
EINECS:248-083-4
UN:2811
ADR:6.1,III
CAS Number:26878-35-3
Formula:C₅H₉Cl₂N₃
MDL Number:MFCD00040818