2,5-Diaminopyridine dihydrochloride, Sigma-Aldrich®
Catalog # SIAL193755-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:182.05 g/mol
Melting Pt:264 °C
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:248-083-4
CAS Number:26878-35-3
Formula:C₅H₉Cl₂N₃
MDL Number:MFCD00040818