Pyridine-2,3-dicarboxylic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:89-00-9
:
Formula:C₇H₅NO₄
MDL Number:MFCD00006295
:167.12 g/mol
Merck Index:12,08258
Boiling Pt:357 °C (1013 hPa)
Melting Pt:188…190 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:201-874-8