Isobutanol
1 mL
Catalog # ACCUAPP-9-120N-1ML
Supplier: AccuStandard
Jump to:
Product Details & Documents
Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
- CAS Number:78-83-1
- Formula:(CH₃)₂CHCH₂OH
- MDL Number:MFCD00004740
- Molecular Weight:74.12 g/mol
- Boiling Pt:65 °C
- Melting Pt:–93.9 °C
- Melting Pt:−93.9 °C
- Flash Pt:52 °F (11 °C) (tcc)
- Storage Temperature:Ambient
- UN:1230
- ADR:3,II
Specifications
Specifications
- Pk:1 mL



