


(S)-(-)-1,2-Epoxypropane ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16088-62-3
:
Formula:C₃H₆O
MDL Number:MFCD00064312
:58.08 g/mol
Merck Index:14,7856
Boiling Pt:34…35 °C
Melting Pt:–112 °C
Density:0.829
Flash Pt:–37 °C (–35 °F)
Storage Temperature:Ambient
EINECS:240-241-0
UN:1280
ADR:3,I
:GHS02
:GHS08
:GHS07