(R)-(+)-1,2-Epoxypropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:15448-47-2
:
Formula:C₃H₆O
MDL Number:MFCD00066211
:58.08 g/mol
Density:0.829 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1280
ADR:3,I
:M-Clarity™ quality level = MQ100