Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,N-Diethyl-p-phenylenediammonium sulphate
N,N-Diethyl-p-phenylenediammonium sulphate 99%
N,N-Diethyl-p-phenylenediammonium sulphate 99%
Irritant
N,N-Diethyl-p-phenylenediammonium sulphate 99%
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Irritant
N,N-Diethyl-p-phenylenediammonium sulphate 99%
Supplier:
Apollo Scientific
New Product
Certificates
About this item
CAS Number:
6283-63-2
:
Formula:
C₁₀H₁₈N₂O₄S
MDL Number:
MFCD00012993
:
262.33 g/mol
Melting Pt:
184...187 °C
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
Documentation