Rotary Vane Pumps
Supplier: Welch by Gardner Denver
Certificates
About this item
Welch Direct-drive vacuum pumps are portable high performance oil-seal pumps. These pumps come with isolation devices integral to the pump to prevent oil and air contamination of your system in the event of a power failure. Pump can be ordered with an optional exhaust fitler to control exhaust oil mist.
- Compact for moving from lab to lab
- Lightweight, compact pump
- Ultimate to 0,004 mbar (<0,003 torr)
- Integral isolation valve