Order Entry
Export
ContactUsLinkComponent
Sigmatrix Urine Diluent
Sigmatrix Urine Diluent
Catalog # SIALSAE0074-1L
Supplier:  Merck
CAS Number:  
Sigmatrix Urine Diluent
Catalog # SIALSAE0074-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 L
  • Description:
    Synthetic negative urine control
  • Storage temperature:
    Stored at 2...8 °C

Specifications

About this item

Sigmatrix urine diluent is a non-biological diluent that mimics human urine.This product can be used as a diluent for sample prep, controls and calibrators and is recommended for LC-MS based applications.

  • Used in enzymatic hydrolysis of drug glucuronides and LC-MS analysis of drugs and drug metabolites
  • The formulation is dye-free

Buffered solution containing calcium chloride, magnesium chloride, potassium chloride, sodium chloride, sodium phosphate, sodium sulfate, urea, and creatinine with sodium azide as a preservative.

Normal precautions exercised in handling laboratory reagents should be followed. Dispose of waste observing all local, state, provincial, or national regulations.

Biological source: Synthetic
Form: Liquid
Shipped in: Wet ice