Orders LinkContactUsLinkComponent
Accessories for Stereomicroscope
Accessories for Stereomicroscope
Supplier:   ZEISS
Accessories for Stereomicroscope
Supplier:  ZEISS
Certificates
Accessories for Stereomicroscope
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Availability
    Price
  • ZEIS4354339401000
    Base 300 for epi-illumination W×D×H 290×353×25 mm stand base with 2× M8 threaded holes and between plate D 120 mm; compatible to T column 350 mm and central column S 490 mm
    Base 300 for epi-illumination W×D×H 290×353×25 mm stand base with 2× M8 threaded holes and between plate D 120 mm; compatible to T column 350 mm and central column S 490 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • ZEIS4351039050000
    Binocular ergo tube S 1,25 × 5 - 45°
    Binocular ergo tube S 1,25 × 5 - 45°
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • ZEIS4354339002000
    Coarse/fine drive, T column 350 mm - lifting range 220 mm, load capacity 10 kg, for stand bases 300/450 - stand base 450 from version -9902, only without nosepiece
    Coarse/fine drive, T column 350 mm - lifting range 220 mm, load capacity 10 kg, for stand bases 300/450 - stand base 450 from version -9902, only without nosepiece
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • ZEIS4351020000000
    Intermediate tube S, fixed 40 mm
    Intermediate tube S, fixed 40 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items