6,7,9,10-Tetrachloroheptadecane
Catalog # EHERA17358850CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Concentration:100 µg/ml
- Pk:1 mL
- Description:6,7,9,10-Tetrachloroheptadecane 100 µg/ml in cyclohexane
- Shipping conditions:20 °C
- Formula:C₁₇H₃₂Cl₄
- Storage temperature:20 °C
- Molecular weight (Mw):378,25
- Form:Single Solution