Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Histopaque®
Histopaque®, sterile filtered, Sigma-Aldrich®
Histopaque®, sterile filtered, Sigma-Aldrich®
:
SIGMA ALDRICH MICROSCOPY
Histopaque®, sterile filtered, Sigma-Aldrich®
:
SIGMA ALDRICH MICROSCOPY
:
:
1,119 g/ml
:
GHS05