Order Entry
Export
ContactUsLinkComponent
Methyl benzenesulphonate
Corrosive
Irritant
Methyl benzenesulphonate
Catalog # 1424112.
Supplier:  USP
CAS Number:  
Corrosive
Irritant
Methyl benzenesulphonate
Catalog # 1424112.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Ampule
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :172.2 g/mol
Flash Pt:143,0 °C (289,4 °F)
UN:3265
ADR:8,II
 :GHS05
 :GHS07
CAS Number:80-18-2
Formula:C₇H₈O₃S
MDL Number:MFCD00014737