Order Entry
Export
Orders LinkContactUsLinkComponent
10XHis azide

10XHis azide

1 mg
Catalog # AATB12642
Supplier:   AAT Bioquest
10XHis azide
Catalog # AATB12642
Supplier:  AAT Bioquest
CAS Number:  
10XHis azide

1 mg

CAS Number:  
Price
556,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

10XHis azide is an excellent building block to make 10XHis conjugates for developing His tag detection probes and purification tools.

  • Stable for 24 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:1641.71 g/mol
Specifications

Specifications

  • Pk:
    1 mg

Specifications