Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polypropylene glycol
Polypropylene glycol, M.W. ∼4,000
Polypropylene glycol, M.W. ∼4,000
Polypropylene glycol, M.W. ∼4,000
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Polypropylene glycol, M.W. ∼4,000
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
25322-69-4
:
Formula:
H(OCH₂CH₂CH₂)nOH
MDL Number:
MFCD00084446
:
1025
Storage Temperature:
Ambient
EINECS:
500-039-8
Related Information
Documentation
Certificates
Search by Lot Number
externalurl.pdf
SDS
msds_ALFA040815
Videos
Thermo Scientific Acroseal 25 Years
Documentation