
Glycolic acid ≥70% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-14-1
:
Formula:C₂H₄O₃
MDL Number:MFCD00004312
:76.05 g/mol
Density:1.262 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-180-5
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ200
:GHS05