Xylitol
Glass bottle for solids, 500 g
Catalog # MOLEM77558672
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
- CAS Number:87-99-0
- Formula:C₅H₁₂O₅
- MDL Number:MFCD00064292
- Molecular Weight:152.15 g/mol
- Merck Index:13,10140
- Boiling Pt:495 °C (1013 hPa)
- Melting Pt:94 °C
- Density:1.52 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-788-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



