Order Entry
Export
Orders LinkContactUsLinkComponent
 

Xylitol

Supplier:   Roth Carl
Xylitol
Supplier:  Roth Carl
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
  • CAS Number:87-99-0
  • Formula:C₅H₁₂O₅
  • MDL Number:MFCD00064292
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,10140
  • Boiling Pt:495 °C (1013 hPa)
  • Melting Pt:94 °C
  • Density:1.52 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-788-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 29,00 €
    29,00 €
    1 * 250 g
       29,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 107,00 €
    107,00 €
    1 * 1 kg
       107,00 €