3,5-Bis(trifluoromethyl)phenylboronic acid, Sigma-Aldrich®
Catalog # SIAL471070-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:257.93 g/mol
CAS Number:73852-19-4
Formula:C₈H₅BF₆O₂
MDL Number:MFCD00051850