Order Entry
Export
Orders LinkContactUsLinkComponent
 

(±)-β-Citronellol 95%

(±)-β-Citronellol 95%
Certificates
(±)-β-Citronellol 95%

3 Options Available Below

Product Details & Documents
Synonyms:  DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol
  • CAS Number:106-22-9
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00002935
  • Molecular Weight:156.27 g/mol
  • Merck Index:13,02354
  • Boiling Pt:222 °C (1013 hPa)
  • Density:0.854 g/cm³
  • Flash Pt:79 °C
  • EINECS:203-375-0
  • ADR:9,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4540 to 1.4580 (20°C, 589 nm)
  • GC:
    ≥94 %
  • Specific gravity:
    (25°C) 0.850 to 0.860
Product Family Options

Product Information