Order Entry
Export
Orders LinkContactUsLinkComponent
 

(±)-β-Citronellol 95%

(±)-β-Citronellol 95%
Certificates
(±)-β-Citronellol 95%
(±)-β-Citronellol 95%

2 Options Available Below

Product Details & Documents

Purity: 95% CAS No.: 106-22-9 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 98°(208°F) Refractive Index: 1.4575 Odour: Pleasant Melting Point: -20° Boiling Point: 221-222° Beilstein Reference: 1721507 Merck Reference: 14,2330 UN No.: 3082 MDL No.: MFCD00002935

Synonyms:  DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol
  • CAS Number:106-22-9
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00002935
  • Molecular Weight:156.27 g/mol
  • Merck Index:14,2330
  • Boiling Pt:221…222 °C
  • Melting Pt:< –20 °C
  • Density:0.858
  • Flash Pt:98 °C (208 °F)
  • Storage Temperature:Ambient
  • EINECS:203-375-0
  • UN:3082
  • ADR:9,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 41,80 €
    41,80 €
    1 * 100 g
       41,80 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 144,00 €
    144,00 €
    1 * 500 g
       144,00 €