Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dibromo-3-pentanone
2,4-Dibromo-3-pentanone, Sigma-Aldrich®
2,4-Dibromo-3-pentanone, Sigma-Aldrich®
2,4-Dibromo-3-pentanone, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2,4-Dibromo-3-pentanone, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
815-60-1
:
Formula:
H₃CBrCHC(O)BrCHCH₃
MDL Number:
MFCD00015719
:
243.93 g/mol
ADR:
8,II
:
M-Clarity™ quality level = MQ100
UN:
3265