Triethylene glycol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-27-6
:
Formula:C₆H₁₄O₄
MDL Number:MFCD00002880
:150.17 g/mol
Merck Index:12,09802
Boiling Pt:285 °C (1013 hPa)
Melting Pt:–7 °C
Density:1.125 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-953-2