Order Entry
Export
ContactUsLinkComponent
Trastuzumab (Human) ELISA Kit
Trastuzumab (Human) ELISA Kit
Catalog # ABNOKA6819
Supplier:  Abnova
New Product
CAS Number:  
Trastuzumab (Human) ELISA Kit
Catalog # ABNOKA6819
Supplier:  Abnova
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Conjugate ELISA:
    HRP
  • Primary antibody reactivity:
    Human
  • Target protein:
    Trastuzumab
  • Description:
    Trastuzumab (Human) ELISA kit (quantitative)
  • Sample type:
    Serum or plasma
  • Detection method:
    Colorimetric (450/650 nm absorbance)
  • Assay Principle:
    Quantitative
  • Detection range:
    10 - 300 ng/ml
  • Storage temperature:
    Store the kit at 4 °C
  • Detection limit:
    3 ng/ml
  • Sample volume:
    10 μl

Specifications

About this item

Trastuzumab (Human) ELISA Kit is a quantitative determination of free Trastuzumab in serum and plasma.

Caution: For research use only (RUO).