1,3-Diphenyl-2-thiourea, Sigma-Aldrich®
Catalog # SIALT28355-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:228.32 g/mol
Merck Index:13,03366
Boiling Pt:365 °C (1013 hPa)
Melting Pt:150…153 °C
Density:1.3205 g/cm³ (20 °C)
EINECS:203-004-2
UN:2811
ADR:6.1,II
:M-Clarity™ quality level = MQ100
CAS Number:102-08-9
Formula:C₁₃H₁₂N₂S
MDL Number:MFCD00004921