di-Sodium succinate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6106-21-4
:
Formula:NaOOCCH₂CH₂COONa·6H₂O
MDL Number:MFCD00149117
:270.14 g/mol
Merck Index:13,08754
Melting Pt:300 °C
Density:1.529 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:205-778-7