D-(-)-Ribose, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-69-1
:
Formula:C₅H₁₀O₅
MDL Number:MFCD00135453
:150.13 g/mol
Merck Index:12,08371
Boiling Pt:331 °C (1013 hPa)
Melting Pt:90…95 °C
Density:0.8 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-059-4