1,8-Naphthalimide, Sigma-Aldrich®
Catalog # SIALN1658-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:197.19 g/mol
Boiling Pt:475 °C (1013 hPa)
Melting Pt:299…300 °C
Density:1.482 g/cm³ (19.85 °C)
:M-Clarity™ quality level = MQ100
EINECS:201-379-7
CAS Number:81-83-4
Formula:C₁₂H₇NO₂
MDL Number:MFCD00006920