Order Entry
Export
Orders LinkContactUsLinkComponent
3-Methylpyrocatechol, Sigma-Aldrich®

3-Methylpyrocatechol, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIALM34006-25G
Supplier:   Merck
3-Methylpyrocatechol, Sigma-Aldrich®
Catalog # SIALM34006-25G
Supplier:  Merck
CAS Number:  488-17-5
3-Methylpyrocatechol, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  488-17-5
Price
125,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Methylcatechol, 3-Methylbenzene-1,2-diol
  • CAS Number:488-17-5
  • Formula:C₇H₈O₂
  • MDL Number:MFCD00016435
  • Molecular Weight:124.14 g/mol
  • Boiling Pt:241 °C (1013 hPa)
  • Melting Pt:58…62 °C
  •  :M-Clarity™ quality level = MQ100
  • EINECS:207-672-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications