2-Mercaptobenzothiazole, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT
- CAS Number:149-30-4
- Formula:C₇H₅NS₂
- MDL Number:MFCD00005781
- Molecular Weight:167.26 g/mol
- Merck Index:13,05893
- Boiling Pt:302 °C (1013 hPa)
- Melting Pt:177…182 °C
- Density:1.42 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-736-8
- ADR:9,III
- :M-Clarity™ quality level = MQ200
- UN:3077
3 Options Available Below
Product Details & Documents
Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT
- CAS Number:149-30-4
- Formula:C₇H₅NS₂
- MDL Number:MFCD00005781
- Molecular Weight:167.26 g/mol
- Merck Index:13,05893
- Boiling Pt:302 °C (1013 hPa)
- Melting Pt:177…182 °C
- Density:1.42 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-736-8
- ADR:9,III
- :M-Clarity™ quality level = MQ200
- UN:3077



