Ethyl-3,4-dihydroxybenzoate, Sigma-Aldrich®
Catalog # SIALE24859-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:182.18 g/mol
Melting Pt:134 °C
:M-Clarity™ quality level = MQ100
EINECS:223-529-0
CAS Number:3943-89-3
Formula:C₉H₁₀O₄
MDL Number:MFCD00002199