
1,4-Dibromobutanedione, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6305-43-7
:
Formula:BrH₂CC(O)C(O)CH₂Br
MDL Number:MFCD00000205
:243.88 g/mol
Boiling Pt:250 °C (1013 hPa)
Melting Pt:117 °C
EINECS:228-615-1
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS07