4,6-Dimethylpyrimidine, Sigma-Aldrich®
Catalog # SIALD183202-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:108.14 g/mol
Boiling Pt:154 °C (1013 hPa)
Melting Pt:25 °C
Density:0.98 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:216-314-8
CAS Number:1558-17-4
Formula:C₆H₈N₂
MDL Number:MFCD00006116