5,6-Dimethylbenzimidazole, Sigma-Aldrich®
Catalog # SIALD147206-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:146.19 g/mol
Merck Index:13,03263
Boiling Pt:377 °C (1013 hPa)
Melting Pt:202 °C
:M-Clarity™ quality level = MQ200
EINECS:209-488-1
CAS Number:582-60-5
Formula:C₉H₁₀N₂
MDL Number:MFCD00005603