Order Entry
Export
ContactUsLinkComponent
1,3-Diethyl-2-thiourea, Sigma-Aldrich®
1,3-Diethyl-2-thiourea, Sigma-Aldrich®
Catalog # SIALD100900-250G
Supplier:  Merck
CAS Number:  
1,3-Diethyl-2-thiourea, Sigma-Aldrich®
Catalog # SIALD100900-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :132.23 g/mol
Boiling Pt:169 °C (1013 hPa)
Melting Pt:75…78 °C
Density:1.118 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Flash Pt:186 °C
EINECS:203-308-5
CAS Number:105-55-5
Formula:C₅H₁₂N₂S
MDL Number:MFCD00004925