Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tri-Calcium phosphate
tri-Calcium phosphate, Sigma-Aldrich®
tri-Calcium phosphate, Sigma-Aldrich®
tri-Calcium phosphate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
tri-Calcium phosphate, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
7758-87-4
:
MFCD00867081
Formula:
Ca₃(PO₄)₂
MDL Number:
MFCD00015984
:
310.18 g/mol
Merck Index:
12,01741
Density:
3.14 g/cm³ (20 °C)
EINECS:
231-840-8