Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-4,6-dimethylpyrimidine
2-Amino-4,6-dimethylpyrimidine, Sigma-Aldrich®
2-Amino-4,6-dimethylpyrimidine, Sigma-Aldrich®
2-Amino-4,6-dimethylpyrimidine, Sigma-Aldrich®
SIALA52005-25G
:
Merck
:
2-Amino-4,6-dimethylpyrimidine, Sigma-Aldrich®
SIALA52005-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
123.16 g/mol
:
152…155 °C
:
M-Clarity™ quality level = MQ100
:
212-181-5
:
767-15-7
:
C₆H₉N₃
:
MFCD00006102