Order Entry
Export
ContactUsLinkComponent
Oil of castor, Sigma-Aldrich®
Oil of castor, Sigma-Aldrich®
Catalog # SIAL83912-1L
Supplier:  Merck
CAS Number:  
Oil of castor, Sigma-Aldrich®
Catalog # SIAL83912-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi
 :
EINECS:232-293-8
CAS Number:8001-79-4
MDL Number:MFCD00130746
Boiling Pt:313
Melting Pt:–10 °C
Density:0.95 g/cm³ (20 °C)
Flash Pt:>113 °C (closed cup)
Merck Index:13,01908
 :M-Clarity™ quality level = MQ100