2,7-Dibromonaphthalene, Sigma-Aldrich®
Catalog # SIAL690171-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:285.97 g/mol
CAS Number:58556-75-5
Formula:C₁₀H₆Br₂
MDL Number:MFCD00238580