Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Diaminophenazine
2,3-Diaminophenazine, Sigma-Aldrich®
2,3-Diaminophenazine, Sigma-Aldrich®
2,3-Diaminophenazine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2,3-Diaminophenazine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
655-86-7
:
Formula:
C₁₂H₁₀N₄
MDL Number:
MFCD00160693
:
210.24 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
211-512-0