5-Bromo-2-iodopyrimidine, Sigma-Aldrich®
Catalog # SIAL637750-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:284.88 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:183438-24-6
Formula:C₄H₂BrIN₂
MDL Number:MFCD01318111