Order Entry
Export
Orders LinkContactUsLinkComponent
3-(4-Bromophenyl)propionic acid, Sigma-Aldrich®

3-(4-Bromophenyl)propionic acid, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIAL595438-5G
Supplier:   Merck
3-(4-Bromophenyl)propionic acid, Sigma-Aldrich®
Catalog # SIAL595438-5G
Supplier:  Merck
CAS Number:  1643-30-7
3-(4-Bromophenyl)propionic acid, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  1643-30-7
Price
74,10 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:1643-30-7
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310793
  • Molecular Weight:229.07 g/mol
  •  :M-Clarity™ quality level = MQ100
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications