Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tri-tert-butylphosphine
Tri-tert-butylphosphine, Sigma-Aldrich®
Tri-tert-butylphosphine, Sigma-Aldrich®
Tri-tert-butylphosphine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Tri-tert-butylphosphine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
13716-12-6
:
Formula:
C₁₂H₂₇P
MDL Number:
MFCD00015006
:
202.32 g/mol
ADR:
4.2,I
:
M-Clarity™ quality level = MQ100
EINECS:
237-266-4
UN:
2846