4,4'-Diaminodiphenyl ether, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:101-80-4
:
Formula:C₁₂H₁₂N₂O
MDL Number:MFCD00007863
:200.24 g/mol
Boiling Pt:389 °C (1013 hPa)
Melting Pt:184 °C
:M-Clarity™ quality level = MQ100
Flash Pt:218 °C
EINECS:202-977-0