Dipotassium hexachloroiridate, Sigma-Aldrich®
Catalog # SIAL450162-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:483.13 g/mol
:M-Clarity™ quality level = MQ100
EINECS:240-976-7
CAS Number:16920-56-2
Formula:Cl₆IrK₂
MDL Number:MFCD00011369